RIE156328709 Rieke Metals, Inc.

3-(3-methylphenyl)-1,1,1-trifluoro-2-propanone

  • MDL number MFCD03844194
  • Cas Number : 898787-61-6
  • Molecular Weight : 202.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6577-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6577-2
Availability details for
Quantity
  • Availability
  • Add to Cart