RIE156328710 Rieke Metals, Inc.

3-(2-Methoxyphenyl)-1,1,1-trifluoro-2-propanone

  • MDL number MFCD03844196
  • Cas Number : 870460-18-7
  • Molecular Weight : 218.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6578-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6578-1
Availability details for
Quantity
  • Availability
  • Add to Cart