RIE156328713 Rieke Metals, Inc.

3-(2-Iodophenyl)-1,1,1-trifluoro-2-propanone

  • MDL number MFCD07700131
  • Cas Number : 898787-65-0
  • Molecular Weight : 314.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6580-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6580-1
Availability details for
Quantity
  • Availability
  • Add to Cart