RIE156328724 Rieke Metals, Inc.

7-(4-Bromophenyl)-7-oxoheptanoic acid

  • MDL number MFCD00671798
  • Cas Number : 898787-83-2
  • Molecular Weight : 299.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7001E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7001E
Availability details for
Quantity
  • Availability
  • Add to Cart