RIE156328726 Rieke Metals, Inc.

6-(3-Bromophenyl)-6-oxohexanoic acid

  • MDL number MFCD00671797
  • Cas Number : 93002-99-4
  • Molecular Weight : 285.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7002D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7002D
Availability details for
Quantity
  • Availability
  • Add to Cart