RIE156328727 Rieke Metals, Inc.

7-(3-bromophenyl)-7-oxoheptanoic acid

  • MDL number MFCD00671800
  • Cas Number : 898787-89-8
  • Molecular Weight : 299.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7002E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7002E
Availability details for
Quantity
  • Availability
  • Add to Cart