RIE156328730 Rieke Metals, Inc.

7-(4-fluorophenyl)-7-oxoheptanoic acid

  • MDL number MFCD01311598
  • Cas Number : 898787-95-6
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7003E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7003E
Availability details for
Quantity
  • Availability
  • Add to Cart