RIE156328732 Rieke Metals, Inc.

6-(4-Chlorophenyl)-6-oxohexanoic acid

  • MDL number MFCD01320020
  • Cas Number : 56721-40-5
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7004D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7004D
Availability details for
Quantity
  • Availability
  • Add to Cart