RIE156328733 Rieke Metals, Inc.

7-(4-Chlorophenyl)-7-oxoheptanoic acid

  • MDL number MFCD01320021
  • Cas Number : 35333-20-1
  • Molecular Weight : 254.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7004E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7004E
Availability details for
Quantity
  • Availability
  • Add to Cart