RIE156328739 Rieke Metals, Inc.

5-(3,5-Ditrifluoromethylphenyl)-5-oxovaleric acid

  • MDL number MFCD01311600
  • Cas Number : 898788-07-3
  • Molecular Weight : 328.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7008C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7008C
Availability details for
Quantity
  • Availability
  • Add to Cart