RIE156328741 Rieke Metals, Inc.

7-(3,5-ditrifluoromethylphenyl)-7-oxoheptanoic acid

  • MDL number MFCD01311602
  • Cas Number : 898788-13-1
  • Molecular Weight : 356.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7008E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7008E
Availability details for
Quantity
  • Availability
  • Add to Cart