RIE156328742 Rieke Metals, Inc.

8-(3,5-ditrifluoromethylphenyl)-8-oxooctanoic acid

  • MDL number MFCD01311601
  • Cas Number : 898788-16-4
  • Molecular Weight : 370.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7008F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7008F
Availability details for
Quantity
  • Availability
  • Add to Cart