RIE156328745 Rieke Metals, Inc.

7-(3,4-Dimethylphenyl)-7-oxoheptanoic acid

  • MDL number MFCD01320030
  • Cas Number : 898788-19-7
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7012E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7012E
Availability details for
Quantity
  • Availability
  • Add to Cart