RIE156328746 Rieke Metals, Inc.

8-(3,4-Dimethylphenyl)-8-oxooctanoic acid

  • MDL number MFCD01320031
  • Cas Number : 898765-12-3
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7012F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7012F
Availability details for
Quantity
  • Availability
  • Add to Cart