RIE156328747 Rieke Metals, Inc.

4-(3,5-Dimethyl-4-methoxyphenyl)-4-oxobutyric acid

  • MDL number MFCD01320032
  • Cas Number : 103234-38-4
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7013B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7013B
Availability details for
Quantity
  • Availability
  • Add to Cart