RIE156328750 Rieke Metals, Inc.

7-(3,5-Dimethyl-4-methoxyphenyl)-7-oxoheptanoic acid

  • MDL number MFCD01320035
  • Cas Number : 898765-15-6
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7013E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7013E
Availability details for
Quantity
  • Availability
  • Add to Cart