RIE156328751 Rieke Metals, Inc.

8-(3,5-Dimethyl-4-methoxyphenyl)-8-oxooctanoic acid

  • MDL number MFCD01320036
  • Cas Number : 898765-18-9
  • Molecular Weight : 292.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7013F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7013F
Availability details for
Quantity
  • Availability
  • Add to Cart