RIE156328754 Rieke Metals, Inc.

6-(2-Bromophenyl)-6-oxohexanoic acid

  • MDL number MFCD00671796
  • Cas Number : 898765-24-7
  • Molecular Weight : 285.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7014D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7014D
Availability details for
Quantity
  • Availability
  • Add to Cart