RIE156328757 Rieke Metals, Inc.

6-(2-methylphenyl)-6-oxohexanoic acid

  • MDL number MFCD01320040
  • Cas Number : 107151-33-7
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7023D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7023D
Availability details for
Quantity
  • Availability
  • Add to Cart