RIE156328758 Rieke Metals, Inc.

7-(2-methylphenyl)-7-oxoheptanoic acid

  • MDL number MFCD01320041
  • Cas Number : 898765-33-8
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7023E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7023E
Availability details for
Quantity
  • Availability
  • Add to Cart