RIE156328760 Rieke Metals, Inc.

6-(3-methylphenyl)-6-oxohexanoic acid

  • MDL number MFCD01320045
  • Cas Number : 56721-44-9
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7027D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7027D
Availability details for
Quantity
  • Availability
  • Add to Cart