RIE156328761 Rieke Metals, Inc.

7-(3-methylphenyl)-7-oxoheptanoic acid

  • MDL number MFCD01320046
  • Cas Number : 898765-39-4
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7027E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7027E
Availability details for
Quantity
  • Availability
  • Add to Cart