RIE156328762 Rieke Metals, Inc.

8-(3-methylphenyl)-8-oxooctanoic acid

  • MDL number MFCD01320047
  • Cas Number : 898765-42-9
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7027F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7027F
Availability details for
Quantity
  • Availability
  • Add to Cart