RIE156328763 Rieke Metals, Inc.

6-(4-Methylphenyl)-6-oxohexanoic acid

  • MDL number MFCD01320048
  • Cas Number : 100847-96-9
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7031D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7031D
Availability details for
Quantity
  • Availability
  • Add to Cart