RIE156328766 Rieke Metals, Inc.

2,2-Dimethyl-4-(3,5-dimethylphenyl)-4-oxobutyric acid

  • MDL number MFCD01320051
  • Cas Number : 898765-45-2
  • Molecular Weight : 234.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7035

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7035
Availability details for
Quantity
  • Availability
  • Add to Cart