RIE156328768 Rieke Metals, Inc.

7-(3,5-dimethylphenyl)-7-oxoheptanoic acid

  • MDL number MFCD01320055
  • Cas Number : 898765-48-5
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7036E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7036E
Availability details for
Quantity
  • Availability
  • Add to Cart