RIE156328771 Rieke Metals, Inc.

7-(3,5-dichlorophenyl)-7-oxoheptanoic acid

  • MDL number MFCD01320060
  • Cas Number : 898765-54-3
  • Molecular Weight : 289.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7038E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7038E
Availability details for
Quantity
  • Availability
  • Add to Cart