RIE156328775 Rieke Metals, Inc.

8-(3-Methoxyphenyl)-8-oxooctanoic acid

  • MDL number MFCD01311620
  • Cas Number : 898765-63-4
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7040F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7040F
Availability details for
Quantity
  • Availability
  • Add to Cart