RIE156328777 Rieke Metals, Inc.

7-(3-fluorophenyl)-7-oxoheptanoic acid

  • MDL number MFCD01311617
  • Cas Number : 898765-67-8
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7042E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7042E
Availability details for
Quantity
  • Availability
  • Add to Cart