RIE156328779 Rieke Metals, Inc.

6-(3-chlorophenyl)-6-oxohexanoic acid

  • MDL number MFCD01320066
  • Cas Number : 898765-71-4
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7044D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7044D
Availability details for
Quantity
  • Availability
  • Add to Cart