RIE156328781 Rieke Metals, Inc.

8-(3-Chlorophenyl)-8-oxooctanoic acid

  • MDL number MFCD01320068
  • Cas Number : 898765-75-8
  • Molecular Weight : 268.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7044F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7044F
Availability details for
Quantity
  • Availability
  • Add to Cart