RIE156328782 Rieke Metals, Inc.

6-(3,4-difluorophenyl)-6-oxohexanoic acid

  • MDL number MFCD01320071
  • Cas Number : 898765-77-0
  • Molecular Weight : 242.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7046D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7046D
Availability details for
Quantity
  • Availability
  • Add to Cart