RIE156328788 Rieke Metals, Inc.

4-Oxo-4-(3,4,5-trifluorophenyl)butyric acid

  • MDL number MFCD01320079
  • Cas Number : 898765-87-2
  • Molecular Weight : 232.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7050B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7050B
Availability details for
Quantity
  • Availability
  • Add to Cart