RIE156328789 Rieke Metals, Inc.

5-Oxo-5-(3,4,5-trifluorophenyl)valeric acid

  • MDL number MFCD01320080
  • Cas Number : 898765-89-4
  • Molecular Weight : 246.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7050C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7050C
Availability details for
Quantity
  • Availability
  • Add to Cart