RIE156328793 Rieke Metals, Inc.

8-oxo-8-(3-phenoxyphenyl)octanoic acid

  • MDL number MFCD02261020
  • Molecular Weight : 326.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7056F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7056F
Availability details for
Quantity
  • Availability
  • Add to Cart