RIE156328794 Rieke Metals, Inc.

6-Oxo-6-(4-thiomethylphenyl)hexanoic acid

  • MDL number MFCD08064682
  • Cas Number : 898765-97-4
  • Molecular Weight : 252.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7057D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7057D
Availability details for
Quantity
  • Availability
  • Add to Cart