RIE156328795 Rieke Metals, Inc.

4-(2-Methoxyphenyl)-4-oxobutyric acid

  • MDL number MFCD01311637
  • Cas Number : 103987-16-2
  • Molecular Weight : 208.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7060B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7060B
Availability details for
Quantity
  • Availability
  • Add to Cart