RIE156328797 Rieke Metals, Inc.

6-(2-methoxyphenyl)-6-oxohexanoic acid

  • MDL number MFCD01311638
  • Cas Number : 107151-39-3
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7060D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7060D
Availability details for
Quantity
  • Availability
  • Add to Cart