RIE156328800 Rieke Metals, Inc.

7-(1-naphthyl)-7-oxoheptanoic acid

  • MDL number MFCD01320089
  • Cas Number : 13672-47-4
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7061E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7061E
Availability details for
Quantity
  • Availability
  • Add to Cart