RIE156328802 Rieke Metals, Inc.

6-(2-Naphthyl)-6-oxohexanoic acid

  • MDL number MFCD01320091
  • Cas Number : 132104-10-0
  • Molecular Weight : 256.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7062D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7062D
Availability details for
Quantity
  • Availability
  • Add to Cart