RIE156328803 Rieke Metals, Inc.

4-oxo-4-(9-Phenanthryl)butyric acid

  • MDL number MFCD01320093
  • Cas Number : 68151-15-5
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7063B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7063B
Availability details for
Quantity
  • Availability
  • Add to Cart