RIE156328804 Rieke Metals, Inc.

5-oxo-5-(9-Phenanthryl)valeric acid

  • MDL number MFCD01320094
  • Cas Number : 898766-03-5
  • Molecular Weight : 292.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7063C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7063C
Availability details for
Quantity
  • Availability
  • Add to Cart