RIE156328806 Rieke Metals, Inc.

7-oxo-7-(9-Phenanthryl)heptanoic acid

  • MDL number MFCD02261199
  • Cas Number : 898766-07-9
  • Molecular Weight : 320.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7063E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7063E
Availability details for
Quantity
  • Availability
  • Add to Cart