RIE156328807 Rieke Metals, Inc.

8-oxo-8-(9-Phenanthryl)octanoic acid

  • MDL number MFCD02261200
  • Cas Number : 898766-09-1
  • Molecular Weight : 334.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7063F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7063F
Availability details for
Quantity
  • Availability
  • Add to Cart