RIE156328808 Rieke Metals, Inc.

4-(2,3-Difluorophenyl)-4-oxobutyric acid

  • MDL number MFCD01320100
  • Cas Number : 898766-11-5
  • Molecular Weight : 214.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7065B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7065B
Availability details for
Quantity
  • Availability
  • Add to Cart