RIE156328809 Rieke Metals, Inc.

5-(2,3-difluorophenyl)-5-oxovaleric acid

  • MDL number MFCD01320101
  • Cas Number : 898766-13-7
  • Molecular Weight : 228.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7065C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7065C
Availability details for
Quantity
  • Availability
  • Add to Cart