RIE156328814 Rieke Metals, Inc.

6-(2,4-difluorophenyl)-6-oxohexanoic acid

  • MDL number MFCD01320105
  • Molecular Weight : 242.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7066D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7066D
Availability details for
Quantity
  • Availability
  • Add to Cart