RIE156328815 Rieke Metals, Inc.

7-(2,4-difluorophenyl)-7-oxoheptanoic acid

  • MDL number MFCD01320106
  • Cas Number : 898766-28-4
  • Molecular Weight : 256.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7066E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7066E
Availability details for
Quantity
  • Availability
  • Add to Cart