RIE156328818 Rieke Metals, Inc.

6-(2,5-difluorophenyl)-6-oxohexanoic acid

  • MDL number MFCD01320109
  • Cas Number : 871127-77-4
  • Molecular Weight : 242.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7067D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7067D
Availability details for
Quantity
  • Availability
  • Add to Cart