RIE156328819 Rieke Metals, Inc.

7-(2,5-difluorophenyl)-7-oxoheptanoic acid

  • MDL number MFCD01320110
  • Cas Number : 898766-37-5
  • Molecular Weight : 256.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7067E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7067E
Availability details for
Quantity
  • Availability
  • Add to Cart